Learn More
Sino Biological Mouse IL4R Protein 1MG (Sino Biological)
Supplier:  Sino Biological 51180M02H1
Encompass_Preferred
A DNA sequence encoding the mouse IL4R (AAB87750.1) (Met1-Arg233) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-237-3863