Learn More
Sino Biological Anti-IL-5 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 51068T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse IL-5 / IL5 / Interleukin 5 (rM IL-5 / IL5 / Interleukin 5; Catalog#51068-M08B; P04401; Met1-Gly133). IL-5 / IL5 / Interleukin 5 specific IgG was purified by Mouse IL-5 / IL5 / Interleukin 5 affinity chromatography.
Catalog No. 50-241-1240