Learn More
Sino Biological Recombinant Mouse IL-5 Protein (His Tag)
Supplier:  Sino Biological 51068M08B1
Encompass_Preferred
A DNA sequence encoding the mouse IL5 (P04401) (Met 1-Gly 133) was expressed,with a C-terminal polyhistidine tag.
Catalog No. 50-255-3015