Learn More
Sino Biological Anti-IL-6 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50136RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse IL6 / IL-6 (Catalog#50136-MNAE; P08505; Phe25-Thr211). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-241-1904