Learn More
Sino Biological Mouse IL6 PAb 100ul (Sino Biological)
Supplier:  Sino Biological 50136T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse IL6/IL-6/Interleukin-6 (rM IL6/IL-6/Interleukin-6; Catalog#50136-MNAE; P08505; Phe25-Thr211). IL6/IL-6/Interleukin-6 specific IgG was purified by Mouse IL6/IL-6/Interleukin-6 affinity chromatography.
Catalog No. 50-237-3128