Learn More
Sino Biological Mouse IL6ST ELISA Pair Set 15PP (Sino Biological)
Catalog No. 502372835
Encompass_Preferred
Each vial contains 60 ng of recombinant mouse IL6ST. Reconstitute with 1 mL detection antibody dilution buffer. After reconstitution, store at -20℃ to -80℃ in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 750 pg/mL is recommended.
Catalog No. 50-237-2835 Supplier Sino Biological Supplier No. SEK5013515
Each vial contains 60 ng of recombinant mouse IL6ST. Reconstitute with 1 mL detection antibody dilution buffer. After reconstitution, store at -20℃ to -80℃ in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 750 pg/mL is recommended.