Learn More
Sino Biological Recombinant Mouse gp130/IL6ST Protein (His Tag), HPLC-verified 20
Supplier:  Sino Biological 50135M08H20
Encompass_Preferred
A DNA sequence encoding the mouse gp130 (NP_034690.3) extracellular domain (Met 1-Glu 617) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-224-6603