Learn More
Sino Biological Recombinant Anti-gp130/IL6ST Antibody (APC), Rabbit Monoclonal
Supplier:  Sino Biological 50135R022A25
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse IL6ST/gp130/CD130 (rM IL6ST/gp130/CD130; Catalog#50135-M08H; NP_034690.3; Met1-Glu617) and conjugated with APC under optimum conditions, the unreacted APC was removed.
Catalog No. 50-255-3430