Learn More
Sino Biological Anti-gp130/IL6ST Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50135RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse gp130 / CD130 / IL6ST (rM gp130 / CD130 / IL6ST; Catalog#50135-M08H; NP_034690.3; Met 1-Glu 617). gp130 / CD130 / IL6ST specific IgG was purified by Mouse gp130 / CD130 / IL6ST affinity chromatography.
Catalog No. 50-255-3422