Learn More
Sino Biological Recombinant Mouse IL-7 Protein
Supplier:  Sino Biological 50217MNAE1
Encompass_Preferred
A DNA sequence encoding the mouse IL7 (NP_032397.1) (Glu26-Ile154) was expressed and purified with an initial Met.
Catalog No. 50-224-7132