Learn More
Sino Biological Mouse IL7R MAb 100ul (Sino Biological)
Supplier:  Sino Biological 50090R201100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse IL7R alpha (rM IL7R alpha; Catalog#50090-M08H; NP_032398.3; Met1-Asp239).
Catalog No. 50-237-2650