Learn More
Sino Biological Recombinant Mouse IL-9 Protein (His Tag) 500
Supplier:  Sino Biological 51302M08B500
Encompass_Preferred
A DNA sequence encoding the mouse IL9 (NP_032399.1) (Met1-Pro144) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-224-7123