Learn More
Biomatik Corporation Mouse Immunoglobulin E (IgE) CLIA Kit
Supplier:  Biomatik Corporation EKU08880
Encompass_Preferred
Mouse Immunoglobulin E (IgE) CLIA Kit for detecting IgE in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.14-100ng/ml. Sensitivity: 0.05ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P06336. Target Synonyms: IGHE; Immunoglobulin Heavy Constant Epsilon; Ig epsilon chain C region. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8606