Learn More
Biomatik Corporation Mouse Immunoglobulin M (IgM) CLIA Kit
Supplier:  Biomatik Corporation EKU08879
Encompass_Preferred
Mouse Immunoglobulin M (IgM) CLIA Kit for detecting IgM in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.55-400ng/ml. Sensitivity: 0.21ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P01872. Target Synonyms: IGHM; Immunoglobulin Heavy Constant Mu; Ig mu chain C region. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4547