Learn More
Biomatik Corporation Mouse Indoleamine-2,3-Dioxygenase 2 (IDO2) ELISA Kit
Supplier:  Biomatik Corporation EKU04919
Encompass_Preferred
Mouse Indoleamine-2,3-Dioxygenase 2 (IDO2) ELISA Kit for detecting IDO2 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 31.25-2000pg/ml. Sensitivity: 12.9pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q8R0V5. Gene ID: 209176. Target Synonyms: INDOL1; Indoleamine-Pyrrole 2,3 Dioxygenase-Like 1; Indoleamine 2,3-dioxygenase-like protein 1; Indoleamine-pyrrole 2,3-dioxygenase-like protein 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5490