Learn More
Biomatik Corporation Mouse Induced Myeloid Leukemia Cell Differentiation Protein Mcl-1 (MCL1) ELISA Kit
Supplier:  Biomatik Corporation EKU06058
Encompass_Preferred
Mouse Induced Myeloid Leukemia Cell Differentiation Protein Mcl-1 (MCL1) ELISA Kit for detecting MCL1 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.063ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P97287. Gene ID: 17210. Target Synonyms: EAT; MCL1L; MCL1S; BCL2L3; TM; Myeloid Cell Leukemia Sequence 1, Bcl2 Related; Bcl-2-like protein 3; Bcl-2-related protein EAT/mcl1; mcl1/EAT. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5683