Learn More
American Research Products Inc Mouse inducible nitric oXide s
Catalog No. 5000329107
Encompass
Mouse inducible nitric oXide synthase iNOS ELISA KIT from CUSABIO Detection Range 7 8 IU/mL-500 IU/mL Sensitivity 1 95 IU/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich
Catalog No. 50-003-29107 Supplier American Research Products Inc Supplier No. CSBE08326M
Mouse inducible nitric oXide synthase iNOS ELISA KIT from CUSABIO Detection Range 7 8 IU/mL-500 IU/mL Sensitivity 1 95 IU/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich