Learn More
Biomatik Corporation Mouse Inhibin Alpha (INHa) CLIA Kit
Supplier:  Biomatik Corporation EKU11557
Encompass_Preferred
Mouse Inhibin Alpha (INHa) CLIA Kit for detecting INHa in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 12.35-1,000pg/ml. Sensitivity: 5.11pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Chemiluminescence. Assay Time: 2h. Accession Number: Q04997. Gene ID: 16322. Target Synonyms: INH-A. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8124