Learn More
Biomatik Corporation Mouse Inhibin B (INHB) CLIA Kit
Supplier:  Biomatik Corporation EKU11561
Encompass_Preferred
Mouse Inhibin B (INHB) CLIA Kit for detecting INHB in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 3.9-1,000pg/ml. Sensitivity: 1.79pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Chemiluminescence. Assay Time: 2h. Accession Number: Q04999. Gene ID: 16324. Target Synonyms: INH-B. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6172