Learn More
Biomatik Corporation Mouse Inhibitor Of Kappa-Light Polypeptide Gene Enhancer In B-Cells Kinase Beta (IkBKb) ELISA Kit
Supplier:  Biomatik Corporation EKU04944
Encompass_Preferred
Mouse Inhibitor Of Kappa-Light Polypeptide Gene Enhancer In B-Cells Kinase Beta (IkBKb) ELISA Kit for detecting IkBKb in serum, plasma, tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.054ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O88351. Gene ID: 16150. Target Synonyms: IKK2; NFKBIKB; IKK-beta; IKKB; I-kappa-B kinase 2; Nuclear factor NF-kappa-B inhibitor kinase beta. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4735