Learn More
Biomatik Corporation Mouse Inosine Triphosphatase Pyrophosphatase (ITPA) ELISA Kit
Supplier:  Biomatik Corporation EKU04948
Encompass_Preferred
Mouse Inosine Triphosphatase Pyrophosphatase (ITPA) ELISA Kit for detecting ITPA in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.049ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9D892. Gene ID: 16434. Target Synonyms: ITPase; Nucleoside Triphosphate Pyrophosphatase; Inosine Triphosphate; Non-canonical purine NTP pyrophosphatase; Nucleoside-triphosphate diphosphatase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4454