Learn More
American Research Products Inc Mouse inositol 1 4 5 trisp
Catalog No. 5000329253
Encompass
Mouse inositol 1 4 5 -trisphosphate (IP3) ELISA Kit from CUSABIO Detection Range 0 78 pg/mL-50 pg/mL Sensitivity 0 195 pg/mL Sample serum plasma cell culture supernates tissue homogenates cell lysates Method Sandwich
Catalog No. 50-003-29253 Supplier American Research Products Inc Supplier No. CSBE13410M
Mouse inositol 1 4 5 -trisphosphate (IP3) ELISA Kit from CUSABIO Detection Range 0 78 pg/mL-50 pg/mL Sensitivity 0 195 pg/mL Sample serum plasma cell culture supernates tissue homogenates cell lysates Method Sandwich