Learn More
Biomatik Corporation Mouse Inositol Polyphosphate-4-Phosphatase Type I 107kDa (INPP4A) ELISA Kit
Supplier:  Biomatik Corporation EKU04950
Encompass_Preferred
Mouse Inositol Polyphosphate-4-Phosphatase Type I 107kDa (INPP4A) ELISA Kit for detecting INPP4A in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.054ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9EPW0. Gene ID: 269180. Target Synonyms: INPP4; Inositol polyphosphate 4-phosphatase type I; Type I inositol 3,4-bisphosphate 4-phosphatase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5193