Learn More
American Research Products Inc Mouse Insulin like growth fact
Supplier:  American Research Products Inc CSBE04584M
Encompass
Mouse Insulin-like growth factor 2 IGF-2 ELISA Kit from CUSABIO Detection Range 31 25 pg/mL-2000 pg/mL Sensitivity 7 8 pg/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich
Catalog No. 50-003-26319