Learn More
American Research Products Inc Mouse Insulin like growth fact
Catalog No. 5000328768
Encompass
Mouse Insulin-like growth factor binding protein 2 IGFBP-2 ELISA KIT from CUSABIO Detection Range 0 45 ng/mL-30 ng/mL Sensitivity 0 11 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-28768 Supplier American Research Products Inc Supplier No. CSBE04589M
Mouse Insulin-like growth factor binding protein 2 IGFBP-2 ELISA KIT from CUSABIO Detection Range 0 45 ng/mL-30 ng/mL Sensitivity 0 11 ng/mL Sample serum plasma tissue homogenates Method Sandwich