Learn More
American Research Products Inc Mouse Insulin like growth fact
Supplier: American Research Products Inc CSBE04587M
Mouse Insulin-like growth factor binding protein 1 IGFBP-1 ELISA Kit from CUSABIO Detection Range 0 312 ng/mL-20 ng/mL Sensitivity 0 078 ng/mL Sample serum plasma tissue homogenates Method Sandwich