Learn More
American Research Products Inc Mouse Insulin like growth fact
Catalog No. 5000330024
Encompass
Mouse Insulin-like growth factor binding protein 3 IGFBP-3 ELISA KIT from CUSABIO Detection Range 0 391 ng/mL-25 ng/mL Sensitivity 0 196 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-30024 Supplier American Research Products Inc Supplier No. CSBE04591M
Mouse Insulin-like growth factor binding protein 3 IGFBP-3 ELISA KIT from CUSABIO Detection Range 0 391 ng/mL-25 ng/mL Sensitivity 0 196 ng/mL Sample serum plasma tissue homogenates Method Sandwich