Learn More
American Research Products Inc Mouse Insulin like growth fact
Supplier:  American Research Products Inc CSBE07355M
Encompass
Mouse Insulin-like growth factor binding protein 4 IGFBP-4 ELISA kit from CUSABIO Detection Range 0 78 ng/mL-50 ng/mL Sensitivity 0 195 ng/mL Sample serum plasma Method Sandwich
Catalog No. 50-003-26782