Learn More
American Research Products Inc Mouse Insulin like growth fact
Supplier:  American Research Products Inc CSBEL011099MO
Encompass
Mouse Insulin-like growth factor-binding protein 5 (IGFBP5) ELISA kit from CUSABIO Detection Range 78 pg/ml -5000pg/ml Sensitivity 19 5 pg/ml Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-27713