Learn More
Biomatik Corporation Mouse Insulin Like Growth Factor Binding Protein 2 (IGFBP2) ELISA Kit
Supplier:  Biomatik Corporation EKU11004
Encompass_Preferred
Mouse Insulin Like Growth Factor Binding Protein 2 (IGFBP2) ELISA Kit for detecting IGFBP2 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 62.5-4000pg/ml. Sensitivity: 23.9pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P47877. Gene ID: 16008. Target Synonyms: IBP2; IGF-BP53. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7145