Learn More
Biomatik Corporation Mouse Insulin Receptor (Anti-INSR) Mini sample ELISA Kit
Supplier:  Biomatik Corporation EKU11629
Encompass_Preferred
Mouse Insulin Receptor (Anti-INSR) Mini sample ELISA Kit for detecting Anti-INSR in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.117ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P15208. Gene ID: 16337. Target Synonyms: CD220; ISR; HHF5; IR. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8116