Learn More
Biomatik Corporation Mouse Integrin Alpha 1 (ITGa1) ELISA Kit
Supplier:  Biomatik Corporation EKU05027
Encompass_Preferred
Mouse Integrin Alpha 1 (ITGa1) ELISA Kit for detecting ITGa1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 78.12-5000pg/ml. Sensitivity: 31pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q3V3R4. Gene ID: 109700. Target Synonyms: CD49-A; CD49a; ITG-A1; VLA1; CD49 antigen-like family member A; Laminin and collagen receptor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5799