Learn More
ACROBiosystems ACROBiosystems Mouse Integrin alpha 11 beta 1 (ITGA11&ITGB1) Heterodimer Protein, His Tag&Tag Free
Supplier:  ACROBiosystems IT1M52W5100UG
Encompass_Preferred
Recombinant Protein;100UG;Mouse ITGA11&ITGB1 Heterodimer Protein, His Tag&Tag Free (IT1-M52W5) is expressed from human 293 cells (HEK293). It contains AA Phe 23 - Pro 1141 (ITGA11) & Gln 21 - Asp 728 (ITGB1) (Accession # P61622-1 (ITGA11) & P09055-1 (ITGB1)).;Mouse ITGA11&ITGB1 Heterodimer Protein, His Tag&Tag Free, produced by co-expression of ITGA11 and ITGB1, has a calculated MW of 132.0 kDa (ITGA11) and 83.5 kDa (ITGB1). Subunit ITGA11 is fused with an acidic tail at the C-terminus and followed by a polyhistidine tag and subunit ITGB1 contains no tag but a basic tail at the C-terminus. The reducing (R) protein migrates as 150-180 kDa due to glycosylation.
Catalog No. 50-210-6783