Learn More
ACROBiosystems ACROBiosystems Mouse Integrin alpha 2 beta 1 (ITGA2&ITGB1) Heterodimer Protein, His Tag&Tag Free (MALS verified)
Catalog No. 502107128
Encompass_Preferred
Recombinant Protein;100UG;Mouse ITGA2&ITGB1 Heterodimer Protein, His Tag&Tag Free (IT1-M52W3) is expressed from human 293 cells (HEK293). It contains AA Tyr 27 - Thr 1129 (ITGA2) & Gln 21 - Asp 728 (ITGB1) (Accession # Q62469-1 (ITGA2) & P09055-1 (ITGB1)).;Mouse ITGA2&ITGB1 Heterodimer Protein, His Tag&Tag Free, produced by co-expression of ITGA2 and ITGB1, has a calculated MW of 127.6 kDa (ITGA2) and 83.5 kDa (ITGB1). Subunit ITGA2 is fused with an acidic tail at the C-terminus and followed by a polyhistidine tag and subunit ITGB1 contains no tag but a basic tail at the C-terminus. The reducing (R) protein migrates as 140-150 kDa (ITGA2) and 100-130 kDa (ITGB1) respectively due to glycosylation.;Immobilized Human Collagen IV at 2 μg/mL (100 μL/well) can bind Mouse ITGA2&ITGB1 Heterodimer Protein, His Tag&Tag Free (Cat. No. IT1-M52W3) with a linear range of 0.02-0.625 μg/mL (QC tested).
Catalog No. 50-210-7128 Supplier ACROBiosystems Supplier No. IT1M52W3100UG
Recombinant Protein;100UG;Mouse ITGA2&ITGB1 Heterodimer Protein, His Tag&Tag Free (IT1-M52W3) is expressed from human 293 cells (HEK293). It contains AA Tyr 27 - Thr 1129 (ITGA2) & Gln 21 - Asp 728 (ITGB1) (Accession # Q62469-1 (ITGA2) & P09055-1 (ITGB1)).;Mouse ITGA2&ITGB1 Heterodimer Protein, His Tag&Tag Free, produced by co-expression of ITGA2 and ITGB1, has a calculated MW of 127.6 kDa (ITGA2) and 83.5 kDa (ITGB1). Subunit ITGA2 is fused with an acidic tail at the C-terminus and followed by a polyhistidine tag and subunit ITGB1 contains no tag but a basic tail at the C-terminus. The reducing (R) protein migrates as 140-150 kDa (ITGA2) and 100-130 kDa (ITGB1) respectively due to glycosylation.;Immobilized Human Collagen IV at 2 μg/mL (100 μL/well) can bind Mouse ITGA2&ITGB1 Heterodimer Protein, His Tag&Tag Free (Cat. No. IT1-M52W3) with a linear range of 0.02-0.625 μg/mL (QC tested).