Learn More
Biomatik Corporation Mouse Integrin Alpha 4 (ITGa4) ELISA Kit
Supplier:  Biomatik Corporation EKU05033
Encompass_Preferred
Mouse Integrin Alpha 4 (ITGa4) ELISA Kit for detecting ITGa4 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.058ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q00651. Gene ID: 16401. Target Synonyms: CD49d; CD49-D; ITG-A4; IA4; Alpha 4 Subunit Of VLA-4 Receptor; CD49 antigen-like family member D; Integrin alpha-IV. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8922