Learn More
ACROBiosystems Mouse Integrin Alpha E Beta 7 Heterodimer Protein, His-Tagged/Tag-Free, 50 μg (Lyophilized)
Catalog No. NC2893265
Encompass_Preferred
Recombinant mouse integrin alpha E beta 7 (ITGAE & ITGB7) heterodimer protein supplied lyophilized in a stabilizing buffer. ITGAE carries a His tag while ITGB7 is tag-free; the protein is expressed in CHO cells and shows >90% purity by SDS-PAGE. Reconstitution instructions are provided in the certificate of analysis.
  • Lyophilized format, 50 μg per vial.
  • His tag on ITGAE subunit; ITGB7 subunit tag-free.
  • Expressed in CHO cells.
  • Purity >90% by SDS-PAGE.
  • Endotoxin <1.0 EU/μg (LAL/rFC).
  • Stable 12 months lyophilized at -20°C to -70°C.
  • Glycosylated; apparent masses ~130-150 kDa (ITGAE) and 90-95 kDa (ITGB7).
Catalog No. NC2893265 Supplier ACROBiosystems Supplier No. IT7M53W350UG
Recombinant mouse integrin alpha E beta 7 (ITGAE & ITGB7) heterodimer protein supplied lyophilized in a stabilizing buffer. ITGAE carries a His tag while ITGB7 is tag-free; the protein is expressed in CHO cells and shows >90% purity by SDS-PAGE. Reconstitution instructions are provided in the certificate of analysis.
  • Lyophilized format, 50 μg per vial.
  • His tag on ITGAE subunit; ITGB7 subunit tag-free.
  • Expressed in CHO cells.
  • Purity >90% by SDS-PAGE.
  • Endotoxin <1.0 EU/μg (LAL/rFC).
  • Stable 12 months lyophilized at -20°C to -70°C.
  • Glycosylated; apparent masses ~130-150 kDa (ITGAE) and 90-95 kDa (ITGB7).