Learn More
Biomatik Corporation Mouse Inter Alpha-Globulin Inhibitor H2 (ITIH2) ELISA Kit
Supplier:  Biomatik Corporation EKU11800
Encompass_Preferred
Mouse Inter Alpha-Globulin Inhibitor H2 (ITIH2) ELISA Kit for detecting ITIH2 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 5.9pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q61703. Gene ID: 16425. Target Synonyms: H2P; SHAP; IGHEP2; Serum-derived hyaluronan-associated protein; Inter-alpha-trypsin inhibitor complex component II. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8080