Learn More
Biomatik Corporation Mouse Intercellular Adhesion Molecule 1 (ICAM1) CLIA Kit
Supplier:  Biomatik Corporation EKU08886
Encompass_Preferred
Mouse Intercellular Adhesion Molecule 1 (ICAM1) CLIA Kit for detecting ICAM1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 68.6-50,000pg/ml. Sensitivity: 29.2pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P13597. Gene ID: 15894. Target Synonyms: CD54; BB2; P3.58; Human Rhinovirus Receptor; Major group rhinovirus receptor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4551