Learn More
Biomatik Corporation Mouse Intercellular Adhesion Molecule 2 (ICAM2) ELISA Kit
Supplier:  Biomatik Corporation EKU05065
Encompass_Preferred
Mouse Intercellular Adhesion Molecule 2 (ICAM2) ELISA Kit for detecting ICAM2 in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.071ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P35330. Gene ID: 15896. Target Synonyms: CD102. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5788