Learn More
Biomatik Corporation Mouse Interferon Alpha 11 (IFNa11) ELISA Kit
Supplier:  Biomatik Corporation EKU11011
Encompass_Preferred
Mouse Interferon Alpha 11 (IFNa11) ELISA Kit for detecting IFNa11 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.3pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q61716. Gene ID: 15964. Target Synonyms: Limitin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8247