Learn More
American Research Products Inc Mouse Interferon b IFN b IFNB
Supplier:  American Research Products Inc CSBE04945MIS
Encompass
Mouse Interferon b IFN-b/IFNB ELISA Kit from CUSABIO Detection Range 31 2pg/mL-1000pg/mL Sensitivity 15 6pg/mL Sample serum plasma Method Sandwich
Catalog No. 50-003-29088