Learn More
American Research Products Inc Mouse Interferon b IFN b IFNB
Catalog No. 5000329378
Encompass
Mouse Interferon b IFN-b/IFNB ELISA Kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma cell culture supernates tissue homogenates cell lysates Method Sandwich
Catalog No. 50-003-29378 Supplier American Research Products Inc Supplier No. CSBE04945M
Mouse Interferon b IFN-b/IFNB ELISA Kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma cell culture supernates tissue homogenates cell lysates Method Sandwich