Learn More
Biomatik Corporation Mouse Interferon Beta (IFNb) Mini sample ELISA Kit
Supplier:  Biomatik Corporation EKU11147
Encompass_Preferred
Mouse Interferon Beta (IFNb) Mini sample ELISA Kit for detecting IFNb in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.8pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P01575. Gene ID: 15977. Target Synonyms: IFNB1; IFN-B; IFB; IFF; IFNB; Interferon Beta 1 Fibroblast. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8211