Learn More
American Research Products Inc Mouse Interferon epsilon IFNE
Catalog No. 5000327707
Encompass
Mouse Interferon epsilon (IFNE) ELISA kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-27707 Supplier American Research Products Inc Supplier No. CSBEL011049MO
Mouse Interferon epsilon (IFNE) ELISA kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma tissue homogenates Method Sandwich