Learn More
American Research Products Inc Mouse interferon inducible pro
Catalog No. 5000326557
Encompass
Mouse interferon-inducible protein 10 IP-10 ELISA Kit from CUSABIO Detection Range 7 8 pg/mL-500 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-26557 Supplier American Research Products Inc Supplier No. CSBE08183MIS
Mouse interferon-inducible protein 10 IP-10 ELISA Kit from CUSABIO Detection Range 7 8 pg/mL-500 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma tissue homogenates Method Sandwich