Learn More
American Research Products Inc Mouse interferon inducible pro
Supplier:  American Research Products Inc CSBE08183M
Encompass
Mouse interferon-inducible protein 10 IP-10 ELISA Kit from CUSABIO Detection Range 0 312 ng/ml - 20 ng/ml Sensitivity 0 078 ng/ml Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-28669