Learn More
Biomatik Corporation Mouse Interferon Inducible T-Cell Alpha Chemoattractant (ITaC) CLIA Kit
Supplier:  Biomatik Corporation EKU09873
Encompass_Preferred
Mouse Interferon Inducible T-Cell Alpha Chemoattractant (ITaC) CLIA Kit for detecting ITaC in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 2.74-2,000pg/ml. Sensitivity: 1.14pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: Q9JHH5. Gene ID: 56066. Target Synonyms: CXCL11; I-TAC; IP9; SCYB11; SCYB9B; B-R1; Chemokine (C-X-C-Motif) ligand 11; Small Inducible Cytokine Subfamily B Member 11; Interferon gamma-inducible protein 9. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8413