Learn More
Biomatik Corporation Mouse Interferon Regulatory Factor 4 (IRF4) ELISA Kit
Supplier:  Biomatik Corporation EKU12494
Encompass_Preferred
Mouse Interferon Regulatory Factor 4 (IRF4) ELISA Kit for detecting IRF4 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q64287. Gene ID: 16364. Target Synonyms: LSIRF; MUM1; NF-EM5; Lymphocyte-specific interferon regulatory factor; Multiple myeloma oncogene 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-7954