Learn More
Cloud Clone Corp Mouse Interleukin 1 Beta (IL1b) (Sandwich ELISA) ELISA Kit
Catalog No. NC4011097
Encompass_Preferred
Mouse Interleukin 1 Beta (IL1b) (Sandwich ELISA) ELISA Kit measures Mouse IL1b in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids.This assay has high sensitivity and excellent specificity for detection of Interleukin 1 Beta (IL1b).No significant cross-reactivity or interference between Interleukin 1 Beta (IL1b) and analogues was observed.Detection Range: 15.6-1,000pg/mLSensitivity:The minimum detectable dose of this kit is typically less than 6.2pg/mLDetection menthod: ColormetricGene ID:16176
Catalog No. NC4011097 Supplier Cloud Clone Corp Supplier No. SEA563MU
Mouse Interleukin 1 Beta (IL1b) (Sandwich ELISA) ELISA Kit measures Mouse IL1b in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids.This assay has high sensitivity and excellent specificity for detection of Interleukin 1 Beta (IL1b).No significant cross-reactivity or interference between Interleukin 1 Beta (IL1b) and analogues was observed.Detection Range: 15.6-1,000pg/mLSensitivity:The minimum detectable dose of this kit is typically less than 6.2pg/mLDetection menthod: ColormetricGene ID:16176