Learn More
American Research Products Inc Mouse Interleukin 1 receptor a
Catalog No. 5000330114
Encompass
Mouse Interleukin 1 receptor antagonist IL-1ra ELISA kit from CUSABIO Detection Range 62 5 pg/mL-4000 pg/mL Sensitivity 15 6 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-30114 Supplier American Research Products Inc Supplier No. CSBE10395M
Mouse Interleukin 1 receptor antagonist IL-1ra ELISA kit from CUSABIO Detection Range 62 5 pg/mL-4000 pg/mL Sensitivity 15 6 pg/mL Sample serum plasma tissue homogenates Method Sandwich